Assay Details
| Gene Symbol: Taco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108302.1 | NP_001101772.1 | 3 - 4 | 708 | 81 | |
| GenBank mRNA | BC166900.1 | - | 3 - 4 | 659 | 6922142 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|

Taco1
Genomic Map