Assay Details
| Gene Symbol: Gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750840.3 | XP_003750888.1 | 4 - 5 | 993 | 166 | |
| GenBank mRNA | BC169088.1 | - | 4 - 5 | 513 | 166 | |
More Information
Set Membership: |
|

Gid4
Genomic Map