Assay Details
| Gene Symbol: Srsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247813.2 | XP_006247875.1 | 2 - 3 | 862 | 103 | |
| GenBank mRNA | FQ232539.1 | - | 2 - 3 | 837 | 103 | |
More Information
Set Membership: |
|

Srsf2
Genomic Map