Assay Details
| Gene Symbol: Spata16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760900.1 | XP_008759122.1 | 2 - 3 | 787 | 56 | |
| XM_008760901.1 | XP_008759123.1 | 2 - 3 | 750 | 56 | ||
More Information
Set Membership: |
|

Spata16
Genomic Map