Assay Details
| Gene Symbol: Ret | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110099.1 | NP_001103569.1 | 18 - 19 | 3055 | 97 | |
| NM_012643.2 | NP_036775.2 | 18 - 19 | 3055 | 97 | ||
| GenBank mRNA | AJ299016.1 | - | 18 - 19 | 3058 | 97 | |
| AJ299017.1 | - | 18 - 19 | 3058 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
ureteric bud development
neural crest cell migration
embryonic epithelial tube formation
activation of cysteine-type endopeptidase activity involved in apoptotic process
homophilic cell adhesion via plasma membrane adhesion molecules
neuron cell-cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
nervous system development
positive regulation of gene expression
positive regulation of neuron projection development
positive regulation of neuron maturation
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron differentiation
positive regulation of cell migration
membrane protein proteolysis
positive regulation of cell adhesion mediated by integrin
ureter maturation
response to drug
neuron maturation
positive regulation of cell size
positive regulation of transcription, DNA-templated
response to pain
enteric nervous system development
regulation of axonogenesis
retina development in camera-type eye
innervation
Peyer's patch morphogenesis
cellular response to retinoic acid
positive regulation of metanephric glomerulus development
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
molecular_function
transmembrane receptor protein tyrosine kinase activity
calcium ion binding
ATP binding
MAPK cascade
ureteric bud development
neural crest cell migration
embryonic epithelial tube formation
activation of cysteine-type endopeptidase activity involved in apoptotic process
homophilic cell adhesion via plasma membrane adhesion molecules
neuron cell-cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
nervous system development
positive regulation of gene expression
positive regulation of neuron projection development
positive regulation of neuron maturation
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
neuron differentiation
positive regulation of cell migration
membrane protein proteolysis
positive regulation of cell adhesion mediated by integrin
ureter maturation
response to drug
neuron maturation
positive regulation of cell size
positive regulation of transcription, DNA-templated
response to pain
enteric nervous system development
regulation of axonogenesis
retina development in camera-type eye
innervation
Peyer's patch morphogenesis
cellular response to retinoic acid
positive regulation of metanephric glomerulus development
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
molecular_function
transmembrane receptor protein tyrosine kinase activity
calcium ion binding
ATP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P480153 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154024 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75450 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480153 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154019 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154024 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480153 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154019 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154024 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P480153 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154019 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154024 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P308662 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480153 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P153972 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701296261 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35495 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55079 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37010 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9197 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701296261 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10864920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864937 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864935 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864936 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864934 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864939 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864927 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864931 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864929 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864933 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864938 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864932 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864922 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10864918 | - | - |
| Rat Genome 230 2.0 Array | 1388208_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1370989_at | - | - |
| Rat Genome 230 2.0 Array | 1370989_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388208_a_at | - | - |
| Rat Genome 230 2.0 Array | 1371112_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371112_at | - | - |
| Affymetrix HT Rat Focus Array | 1388208_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370989_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388208_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370989_at | - | - |
| Affymetrix HT Rat Focus Array | 1371112_at | - | - |
| Affymetrix HT Rat Focus Array | 1371112_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400025355.rn.1 | TC0400003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025356.rn.1 | TC0400003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400025354.rn.1 | TC0400003626.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371112_PM_at | - | - |
| Rat Expression Set 230 | 1388208_a_at | - | - |
| Rat Expression Set 230 | 1388208_PM_a_at | - | - |
| Rat Expression Set 230 | 1370989_at | - | - |
| Rat Expression Set 230 | 1371112_at | - | - |
| Rat Expression Set 230 | 1370989_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10864918 | - | - |
| Rat Genome U34 Set | rc_AI639318_at | - | - |
| Rat Genome U34 Set | rc_AA859878_at | - | - |
| Rat Genome U34 Set | AF042830_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20945440 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372073 | - | - |
| RatRef-12 Expression BeadChip | ILMN_58171 | - | - |

Ret
Genomic Map