Assay Details
| Gene Symbol: Zfp286a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191921.1 | NP_001178850.1 | 3 - 4 | 275 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp286a| Gene Symbol: Zfp286a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191921.1 | NP_001178850.1 | 3 - 4 | 275 | 58 | |
Set Membership: |
|