Assay Details
| Gene Symbol: Gucy2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024380.1 | NP_077356.1 | 5 - 6 | 1532 | 79 | |
| GenBank mRNA | L36029.1 | - | 5 - 6 | 1532 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2e
Genomic Map