Assay Details
| Gene Symbol: Arhgap44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246834.2 | XP_006246896.1 | 19 - 20 | 2260 | 59 | |
| XM_006246835.2 | XP_006246897.1 | 18 - 19 | 2242 | 59 | ||
| XM_008767895.1 | XP_008766117.1 | 19 - 20 | 2260 | 59 | ||
| XM_008767896.1 | XP_008766118.1 | 17 - 18 | 2014 | 59 | ||
More Information
Set Membership: |
|

Arhgap44
Genomic Map