Assay Details
| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053419.1 | NP_445871.1 | 1777 | 76 | ||
| GenBank mRNA | AF169549.1 | - | 748 | 76 | ||
| AF242544.1 | - | 1435 | 76 | |||
| EF121977.1 | - | 1240 | 76 | |||
| EF121978.1 | - | 1240 | 76 | |||
| EF121979.1 | - | 1474 | 76 | |||
| EF121980.1 | - | 1474 | 76 | |||
| EF121981.1 | - | 1777 | 76 | |||
| EF121982.1 | - | 1240 | 76 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P161768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123526 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P161768 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P161768 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P161768 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P177747 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P866704 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52172 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40359 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97892 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10736936 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736943 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736945 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736942 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736935 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736938 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736940 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736937 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736941 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736954 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736956 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736955 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736939 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736949 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10736934 | - | - |
| Rat Genome 230 2.0 Array | 1370717_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388450_at | - | - |
| Rat Genome 230 2.0 Array | 1388450_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370717_at | - | - |
| Affymetrix HT Rat Focus Array | 1370717_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370717_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000040501.rn.1 | TC1000004361.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000040505.rn.1 | TC1000004361.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000040499.rn.1 | TC1000004361.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000040500.rn.1 | TC1000004361.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000040498.rn.1 | TC1000004361.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388450_PM_at | - | - |
| Rat Expression Set 230 | 1388450_at | - | - |
| Rat Expression Set 230 | 1370717_at | - | - |
| Rat Expression Set 230 | 1370717_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10736934 | - | - |
| Rat Genome U34 Set | rc_AI228931_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21238436 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365426 | - | - |

Synrg
Genomic Map