Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191655.1 | NP_001178584.1 | 9 - 10 | 891 | 57 | |
| XM_006247715.2 | XP_006247777.1 | 9 - 10 | 1096 | 57 | ||
| XM_006247716.2 | XP_006247778.1 | 9 - 10 | 942 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem104
Genomic Map