Assay Details
| Gene Symbol: RGD1559732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750838.3 | XP_003750886.1 | 1 - 2 | 239 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003750838.3 transcripts.
Set Membership: |
|

RGD1559732
Genomic Map