Assay Details
| Gene Symbol: RGD1559732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750838.3 | XP_003750886.1 | 4 - 5 | 756 | 80 | |
| XM_006246534.2 | XP_006246596.1 | 3 - 4 | 681 | 80 | ||
More Information
Set Membership: |
|

RGD1559732
Genomic Map