Assay Details
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019206.2 | NP_062079.2 | 8 - 9 | 1155 | 63 | |
XM_008767561.1 | XP_008765783.1 | 8 - 9 | 1159 | 63 | ||
GenBank mRNA | U33472.1 | - | 3 - 4 | 307 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the U33472.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
