Assay Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252375.2 | XP_006252437.1 | 1 - 2 | 3290 | 137 | |
| XM_008770897.1 | XP_008769119.1 | 2 - 3 | 3053 | 137 | ||
More Information
Set Membership: |
|

Pcdh8
Genomic Map