Assay Details
| Gene Symbol: Dgkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252363.2 | XP_006252425.1 | 6 - 7 | 870 | 68 | |
| XM_006252364.2 | XP_006252426.1 | 6 - 7 | 3187 | 68 | ||
| XM_008770918.1 | XP_008769140.1 | 8 - 9 | 1859 | 68 | ||
| XM_008770919.1 | XP_008769141.1 | 8 - 9 | 1858 | 68 | ||
| XM_008770920.1 | XP_008769142.1 | 6 - 7 | 935 | 68 | ||
More Information
Set Membership: |
|

Dgkh
Genomic Map