Assay Details
| Gene Symbol: Rtn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053865.1 | NP_446317.1 | 5 - 6 | 2526 | 88 | |
| XM_008764690.1 | XP_008762912.1 | 3 - 4 | 556 | 88 | ||
| GenBank mRNA | FQ212949.1 | - | 3 - 4 | 467 | 88 | |
| FQ214343.1 | - | 3 - 4 | 392 | 88 | ||
| U17603.1 | - | 3 - 4 | 526 | 88 | ||
| U17604.1 | - | 5 - 6 | 2526 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rtn1
Genomic Map