Assay Details
| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277885.1 | NP_001264814.1 | 1 - 2 | 232 | 125 | |
| XM_008764470.1 | XP_008762692.1 | 1 - 2 | 227 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem247
Genomic Map