Assay Details
| Gene Symbol: RGD1305089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106730.2 | NP_001100200.1 | 6 - 7 | 2133 | 92 | |
| GenBank mRNA | BC168878.1 | - | 6 - 7 | 2133 | 7382020 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P157078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179641 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P157078 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P157078 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P157078 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P157078 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44968 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27807 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76530 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10884651 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884652 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884655 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884650 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884653 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884654 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10884648 | - | - |
| Rat Genome 230 2.0 Array | 1393100_at | - | - |
| Rat Genome 230 2.0 Array | 1374037_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374037_at | - | - |
| Rat Genome 230 2.0 Array | 1393100_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374037_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374037_at | - | - |
| Affymetrix HT Rat Focus Array | 1393100_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393100_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600005299.rn.1 | TC0600000791.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600005300.rn.1 | TC0600000791.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374037_at | - | - |
| Rat Expression Set 230 | 1374037_PM_at | - | - |
| Rat Expression Set 230 | 1393100_at | - | - |
| Rat Expression Set 230 | 1393100_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884648 | - | - |
| Rat Genome U34 Set | rc_AA945739_at | - | - |
| Rat Genome U34 Set | rc_AI013667_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21614436 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530438 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371310 | - | - |

RGD1305089
Genomic Map