Assay Details
| Gene Symbol: Heatr5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240091.1 | XP_006240153.1 | 28 - 29 | 4483 | 68 | |
| XM_006240095.1 | XP_006240157.1 | 27 - 28 | 4456 | 68 | ||
| XM_006240096.1 | XP_006240158.1 | 27 - 28 | 4453 | 68 | ||
| XM_008764667.1 | XP_008762889.1 | 28 - 29 | 4483 | 68 | ||
| XM_008764668.1 | XP_008762890.1 | 28 - 29 | 4480 | 68 | ||
More Information
Set Membership: |
|

Heatr5a
Genomic Map