Assay Details
| Gene Symbol: Pum2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106715.1 | NP_001100185.1 | 7 - 8 | 952 | 85 | |
| XM_006239879.2 | XP_006239941.1 | 7 - 8 | 1030 | 85 | ||
| XM_006239880.2 | XP_006239942.1 | 7 - 8 | 1030 | 85 | ||
| XM_006239883.2 | XP_006239945.1 | 7 - 8 | 1030 | 85 | ||
| XM_006239884.2 | XP_006239946.2 | 6 - 7 | 1706 | 85 | ||
| XM_008764579.1 | XP_008762801.1 | 6 - 7 | 1705 | 85 | ||
| XM_008764580.1 | XP_008762802.1 | 7 - 8 | 1051 | 85 | ||
| XM_008764581.1 | XP_008762803.1 | 6 - 7 | 1705 | 85 | ||
| XM_008764582.1 | XP_008762804.1 | 6 - 7 | 1704 | 85 | ||
| GenBank mRNA | AY724506.1 | - | 7 - 8 | 1011 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109576 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11190 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11190 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11190 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11190 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P340329 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1042901 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P259365 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17033 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18101 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4087 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73325 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42356 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50528 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10883527 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883518 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883525 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883517 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883528 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883520 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883519 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10883510 | - | - |
| Rat Genome 230 2.0 Array | 1382198_at | - | - |
| Rat Genome 230 2.0 Array | 1375534_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398947_at | - | - |
| Rat Genome 230 2.0 Array | 1380604_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375534_at | - | - |
| Rat Genome 230 2.0 Array | 1398947_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382198_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380604_at | - | - |
| Affymetrix HT Rat Focus Array | 1375534_at | - | - |
| Affymetrix HT Rat Focus Array | 1380604_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398947_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1380604_at | - | - |
| Affymetrix HT Rat Focus Array | 1375534_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1398947_at | - | - |
| Affymetrix HT Rat Focus Array | 1382198_at | - | - |
| Affymetrix HT Rat Focus Array | 1382198_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600002688.rn.1 | TC0600000398.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002686.rn.1 | TC0600000398.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002689.rn.1 | TC0600000398.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002691.rn.1 | TC0600000398.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002692.rn.1 | TC0600000398.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002690.rn.1 | TC0600000398.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375534_at | - | - |
| Rat Expression Set 230 | 1398947_at | - | - |
| Rat Expression Set 230 | 1375534_PM_at | - | - |
| Rat Expression Set 230 | 1398947_PM_at | - | - |
| Rat Expression Set 230 | 1382198_at | - | - |
| Rat Expression Set 230 | 1382198_PM_at | - | - |
| Rat Expression Set 230 | 1380604_PM_at | - | - |
| Rat Expression Set 230 | 1380604_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883510 | - | - |
| Rat Genome U34 Set | rc_AA849809_at | - | - |
| Rat Genome U34 Set | rc_AI007781_at | - | - |
| Rat Genome U34 Set | rc_AI072431_at | - | - |
| Rat Genome U34 Set | rc_AI233267_at | - | - |
| Rat Genome U34 Set | rc_AI177894_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21398098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349337 | - | - |

Pum2
Genomic Map