Assay Details
| Gene Symbol: Mrpl33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729600.3 | XP_002729646.1 | 1 - 2 | 99 | 74 | |
| GenBank mRNA | FQ224256.1 | - | 1 - 2 | 60 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002729600.3 transcripts.
Set Membership: |
|

Mrpl33
Genomic Map