Assay Details
| Gene Symbol: Ints2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100630.2 | NP_001094100.2 | 16 - 17 | 2439 | 64 | |
| XM_008768072.1 | XP_008766294.1 | 16 - 17 | 2506 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ints2
Genomic Map