Assay Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019148.2 | NP_062021.1 | 3 - 4 | 462 | 58 | |
| XM_006239622.2 | XP_006239684.1 | 3 - 4 | 558 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Strn
Genomic Map