Assay Details
| Gene Symbol: Etv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108709.3 | NP_001102179.2 | 7 - 8 | 740 | 74 | |
| NM_001163156.3 | NP_001156628.1 | 7 - 8 | 759 | 74 | ||
| XM_006240047.2 | XP_006240109.1 | 6 - 7 | 721 | 74 | ||
| XM_006240048.2 | XP_006240110.1 | 7 - 8 | 779 | 74 | ||
| XM_006240050.2 | XP_006240112.1 | 4 - 5 | 865 | 74 | ||
| XM_008764647.1 | XP_008762869.1 | 6 - 7 | 721 | 74 | ||
| XM_008764648.1 | XP_008762870.1 | 7 - 8 | 1194 | 74 | ||
| GenBank mRNA | BC168860.1 | - | 7 - 8 | 740 | 7121808 | 74 |
| CB718897.1 | - | 1 - 2 | 66 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
axon guidance
muscle organ development
mechanosensory behavior
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
axon guidance
muscle organ development
mechanosensory behavior
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P211061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162617 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P211061 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P211061 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P211061 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P315649 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P211061 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P315647 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10884341 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884344 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884340 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884342 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884346 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884343 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884351 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10884339 | - | - |
| Rat Genome 230 2.0 Array | 1392477_at | - | - |
| Rat Genome 230 2.0 Array | 1382593_at | - | - |
| Rat Genome 230 2.0 Array | 1382593_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392555_at | - | - |
| Rat Genome 230 2.0 Array | 1397015_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392477_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397015_at | - | - |
| Rat Genome 230 2.0 Array | 1392555_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392477_at | - | - |
| Affymetrix HT Rat Focus Array | 1382593_at | - | - |
| Affymetrix HT Rat Focus Array | 1382593_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392477_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600004597.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004610.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004602.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004604.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004598.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004606.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004603.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004608.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004607.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004605.rn.1 | TC0600000653.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600004609.rn.1 | TC0600000653.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382593_at | - | - |
| Rat Expression Set 230 | 1392477_PM_at | - | - |
| Rat Expression Set 230 | 1397015_at | - | - |
| Rat Expression Set 230 | 1392555_at | - | - |
| Rat Expression Set 230 | 1392477_at | - | - |
| Rat Expression Set 230 | 1397015_PM_at | - | - |
| Rat Expression Set 230 | 1392555_PM_at | - | - |
| Rat Expression Set 230 | 1382593_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10884339 | - | - |
| Rat Genome U34 Set | rc_AI059914_at | - | - |
| Rat Genome U34 Set | rc_AA963276_at | - | - |
| Rat Genome U34 Set | rc_AI102948_at | - | - |
| Rat Genome U34 Set | rc_AI231368_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22324791 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373392 | - | - |

Etv1
Genomic Map