Assay Details
| Gene Symbol: Churc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106741.2 | NP_001100211.1 | 1 - 2 | 88 | 83 | |
| GenBank mRNA | BC168770.1 | - | 1 - 2 | 88 | 6918115 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|

Churc1
Genomic Map