Assay Details
| Gene Symbol: Heatr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191064.1 | NP_001177993.1 | 21 - 22 | 3171 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr5b| Gene Symbol: Heatr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191064.1 | NP_001177993.1 | 21 - 22 | 3171 | 62 | |
Set Membership: |
|