Assay Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248413.2 | XP_006248475.1 | 1 - 2 | 124 | 94 | |
| GenBank mRNA | BC097971.1 | - | 1 - 2 | 70 | 7442058 | 94 |
| FQ222091.1 | - | 1 - 2 | 94 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC097971.1,XM_006248413.2 transcripts.
This assay is in the 5'UTR region of the BC097971.1,XM_006248413.2 transcripts.
Set Membership: |
|

Fam162a
Genomic Map