Assay Details
| Gene Symbol: Fam162a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029903.1 | NP_001025074.1 | 2 - 3 | 152 | 62 | |
| XM_006248413.2 | XP_006248475.1 | 2 - 3 | 240 | 62 | ||
| GenBank mRNA | BC097971.1 | - | 2 - 3 | 186 | 7442058 | 62 |
| FQ222091.1 | - | 2 - 3 | 210 | 62 | ||
| M17412.1 | - | 1 - 2 | 125 | 62 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam162a
Genomic Map