Assay Details
Gene Symbol: Muc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107089.2 | NP_001100559.1 | 2 - 3 | 1112 | 68 | |
XM_006248449.2 | XP_006248511.1 | 2 - 3 | 1881 | 68 | ||
GenBank mRNA | BC167011.1 | - | 2 - 3 | 1112 | 7128599 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
