Assay Details
Gene Symbol: Muc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248449.2 | XP_006248511.1 | 1 - 2 | 124 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
