Assay Details
| Gene Symbol: Akap14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021703.1 | NP_067735.1 | 2 - 3 | 1331 | 89 | |
| GenBank mRNA | AJ002474.1 | - | 2 - 3 | 1331 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap14
Genomic Map