Assay Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257472.2 | XP_006257534.1 | 4 - 5 | 939 | 79 | |
| XM_006257473.2 | XP_006257535.1 | 4 - 5 | 555 | 79 | ||
| XM_006257475.1 | XP_006257537.1 | 5 - 6 | 683 | 79 | ||
| XM_008773400.1 | XP_008771622.1 | 5 - 6 | 834 | 79 | ||
| GenBank mRNA | FQ228987.1 | - | 4 - 5 | 573 | 79 | |
| S82627.1 | - | 3 - 4 | 475 | 79 | ||
More Information
Set Membership: |
|

Rhox5
Genomic Map