Assay Details
| Gene Symbol: Klk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107508.1 | NP_001100978.1 | 1 - 2 | 159 | 89 | |
| XM_008759351.1 | XP_008757573.1 | 3 - 4 | 239 | 89 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107508.1 transcripts.
Set Membership: |
|

Klk12
Genomic Map