Assay Details
| Gene Symbol: Apoe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270681.1 | NP_001257610.1 | 2 - 3 | 181 | 94 | |
| NM_001270682.1 | NP_001257611.1 | 2 - 3 | 160 | 94 | ||
| NM_001270683.1 | NP_001257612.1 | 2 - 3 | 147 | 94 | ||
| NM_001270684.1 | NP_001257613.1 | 2 - 3 | 123 | 94 | ||
| NM_138828.3 | NP_620183.2 | 2 - 3 | 141 | 94 | ||
| GenBank mRNA | BC060313.1 | - | 2 - 3 | 114 | 6919048 | 94 |
| BC086581.1 | - | 2 - 3 | 84 | 7307758 | 94 | |
| FQ209421.1 | - | 2 - 3 | 106 | 94 | ||
| FQ209426.1 | - | 2 - 3 | 112 | 94 | ||
| FQ209428.1 | - | 2 - 3 | 110 | 94 | ||
| FQ209457.1 | - | 2 - 3 | 109 | 94 | ||
| FQ209625.1 | - | 2 - 3 | 110 | 94 | ||
| FQ209879.1 | - | 1 - 2 | 57 | 94 | ||
| FQ209926.1 | - | 2 - 3 | 110 | 94 | ||
| FQ210262.1 | - | 2 - 3 | 110 | 94 | ||
| FQ210410.1 | - | 2 - 3 | 110 | 94 | ||
| FQ211250.1 | - | 2 - 3 | 111 | 94 | ||
| FQ212769.1 | - | 2 - 3 | 94 | |||
| FQ213060.1 | - | 2 - 3 | 106 | 94 | ||
| FQ213396.1 | - | 2 - 3 | 106 | 94 | ||
| FQ213533.1 | - | 2 - 3 | 113 | 94 | ||
| FQ213898.1 | - | 2 - 3 | 102 | 94 | ||
| FQ214072.1 | - | 2 - 3 | 110 | 94 | ||
| FQ214282.1 | - | 2 - 3 | 110 | 94 | ||
| FQ218386.1 | - | 2 - 3 | 109 | 94 | ||
| FQ218473.1 | - | 2 - 3 | 139 | 94 | ||
| FQ218485.1 | - | 2 - 3 | 110 | 94 | ||
| FQ218540.1 | - | 2 - 3 | 110 | 94 | ||
| FQ218699.1 | - | 2 - 3 | 112 | 94 | ||
| FQ218898.1 | - | 2 - 3 | 112 | 94 | ||
| FQ218953.1 | - | 1 - 2 | 166 | 94 | ||
| FQ219009.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219124.1 | - | 1 - 2 | 70 | 94 | ||
| FQ219144.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219432.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219448.1 | - | 2 - 3 | 112 | 94 | ||
| FQ219475.1 | - | 2 - 3 | 105 | 94 | ||
| FQ219490.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219516.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219534.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219700.1 | - | 2 - 3 | 110 | 94 | ||
| FQ219711.1 | - | 2 - 3 | 110 | 94 | ||
| FQ220125.1 | - | 2 - 3 | 106 | 94 | ||
| FQ220532.1 | - | 2 - 3 | 106 | 94 | ||
| FQ220683.1 | - | 2 - 3 | 110 | 94 | ||
| FQ220752.1 | - | 2 - 3 | 141 | 94 | ||
| FQ220879.1 | - | 2 - 3 | 114 | 94 | ||
| FQ221856.1 | - | 2 - 3 | 110 | 94 | ||
| FQ223299.1 | - | 1 - 2 | 61 | 94 | ||
| FQ229186.1 | - | 2 - 3 | 110 | 94 | ||
| FQ229201.1 | - | 2 - 3 | 110 | 94 | ||
| FQ229530.1 | - | 1 - 2 | 61 | 94 | ||
| FQ229565.1 | - | 2 - 3 | 106 | 94 | ||
| FQ229650.1 | - | 2 - 3 | 110 | 94 | ||
| FQ229759.1 | - | 2 - 3 | 110 | 94 | ||
| FQ229950.1 | - | 2 - 3 | 110 | 94 | ||
| FQ230084.1 | - | 2 - 3 | 110 | 94 | ||
| FQ231375.1 | - | 2 - 3 | 139 | 94 | ||
| J00705.1 | - | 1 - 2 | 68 | 94 | ||
More Information
Set Membership: |
|

Apoe
Genomic Map