Assay Details
| Gene Symbol: Apoe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138828.3 | NP_620183.2 | 1 - 2 | 74 | 63 | |
| GenBank mRNA | BC060313.1 | - | 1 - 2 | 47 | 6919048 | 63 |
| FQ209421.1 | - | 1 - 2 | 39 | 63 | ||
| FQ209426.1 | - | 1 - 2 | 45 | 63 | ||
| FQ209428.1 | - | 1 - 2 | 43 | 63 | ||
| FQ209625.1 | - | 1 - 2 | 43 | 63 | ||
| FQ210262.1 | - | 1 - 2 | 43 | 63 | ||
| FQ210410.1 | - | 1 - 2 | 43 | 63 | ||
| FQ211250.1 | - | 1 - 2 | 44 | 63 | ||
| FQ212760.1 | - | 1 - 2 | 43 | 63 | ||
| FQ212769.1 | - | 1 - 2 | 43 | 63 | ||
| FQ213060.1 | - | 1 - 2 | 39 | 63 | ||
| FQ213396.1 | - | 1 - 2 | 39 | 63 | ||
| FQ213533.1 | - | 1 - 2 | 46 | 63 | ||
| FQ213898.1 | - | 1 - 2 | 35 | 63 | ||
| FQ214072.1 | - | 1 - 2 | 43 | 63 | ||
| FQ214282.1 | - | 1 - 2 | 43 | 63 | ||
| FQ218386.1 | - | 1 - 2 | 42 | 63 | ||
| FQ218473.1 | - | 1 - 2 | 72 | 63 | ||
| FQ218485.1 | - | 1 - 2 | 43 | 63 | ||
| FQ218699.1 | - | 1 - 2 | 45 | 63 | ||
| FQ218898.1 | - | 1 - 2 | 45 | 63 | ||
| FQ219009.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219144.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219319.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219432.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219448.1 | - | 1 - 2 | 45 | 63 | ||
| FQ219475.1 | - | 1 - 2 | 38 | 63 | ||
| FQ219490.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219516.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219534.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219700.1 | - | 1 - 2 | 43 | 63 | ||
| FQ219711.1 | - | 1 - 2 | 43 | 63 | ||
| FQ220532.1 | - | 1 - 2 | 39 | 63 | ||
| FQ220683.1 | - | 1 - 2 | 43 | 63 | ||
| FQ220752.1 | - | 1 - 2 | 74 | 63 | ||
| FQ220879.1 | - | 1 - 2 | 47 | 63 | ||
| FQ221856.1 | - | 1 - 2 | 43 | 63 | ||
| FQ229201.1 | - | 1 - 2 | 43 | 63 | ||
| FQ229565.1 | - | 1 - 2 | 39 | 63 | ||
| FQ229650.1 | - | 1 - 2 | 43 | 63 | ||
| FQ229759.1 | - | 1 - 2 | 43 | 63 | ||
| FQ229950.1 | - | 1 - 2 | 43 | 63 | ||
| FQ230084.1 | - | 1 - 2 | 43 | 63 | ||
| FQ231375.1 | - | 1 - 2 | 72 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC060313.1,NM_138828.3 transcripts.
Set Membership: |
|

Apoe
Genomic Map