Assay Details
| Gene Symbol: Klk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170405.1 | NP_001163876.1 | 5 - 6 | 835 | 89 | |
| XM_008759331.1 | XP_008757553.1 | 4 - 5 | 754 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klk13
Genomic Map