Assay Details
| Gene Symbol: Zfp111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133323.1 | NP_579857.1 | 1 - 2 | 283 | 148 | |
| GenBank mRNA | U27186.1 | - | 1 - 2 | 283 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133323.1, U27186.1 transcripts.
Set Membership: |
|

Zfp111
Genomic Map