Assay Details
| Gene Symbol: Lin37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106245.1 | NP_001099715.1 | 7 - 8 | 764 | 70 | |
| XM_006228769.2 | XP_006228831.1 | 7 - 8 | 1185 | 70 | ||
| GenBank mRNA | FQ233946.1 | - | 7 - 8 | 732 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lin37
Genomic Map