Assay Details
| Gene Symbol: Apoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109996.1 | NP_001103466.1 | 2 - 3 | 108 | 88 | |
| NM_012824.2 | NP_036956.1 | 1 - 2 | 143 | 88 | ||
| GenBank mRNA | AY327505.1 | - | 2 - 3 | 125 | 88 | |
| BC098670.1 | - | 1 - 2 | 82 | 7372566 | 88 | |
| FQ210440.1 | - | 2 - 3 | 108 | 88 | ||
| FQ210757.1 | - | 2 - 3 | 97 | 88 | ||
| FQ211211.1 | - | 2 - 3 | 109 | 88 | ||
| FQ212377.1 | - | 2 - 3 | 108 | 88 | ||
| FQ223744.1 | - | 2 - 3 | 108 | 88 | ||
| X15512.1 | - | 1 - 2 | 143 | 88 | ||
| Gene Symbol: LOC100911905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752245.2 | XP_003752293.2 | 2 - 3 | 142 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apoc1
Genomic Map