Assay Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127552.1 | NP_001121024.1 | 3 - 4 | 518 | 71 | |
| XM_006228856.2 | XP_006228918.1 | 3 - 4 | 566 | 71 | ||
| XM_006228857.2 | XP_006228919.1 | 3 - 4 | 566 | 71 | ||
| XM_006228858.2 | XP_006228920.1 | 3 - 4 | 566 | 71 | ||
| GenBank mRNA | BC166537.1 | - | 3 - 4 | 518 | 7191211 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Lsm14a
Genomic Map