Assay Details
| Gene Symbol: Fam189a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759569.1 | XP_008757791.1 | 8 - 9 | 1215 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam189a1| Gene Symbol: Fam189a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759569.1 | XP_008757791.1 | 8 - 9 | 1215 | 53 | |
Set Membership: |
|