Assay Details
| Gene Symbol: Dhx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130039.1 | NP_001123511.1 | 8 - 9 | 1970 | 117 | |
| XM_006230443.2 | XP_006230505.1 | 8 - 9 | 2535 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P282843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32251 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P282843 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P282843 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P282843 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10726359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726370 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10711776 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726367 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726368 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726369 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726365 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10711769 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10726358 | - | - |
| Rat Genome 230 2.0 Array | 1395579_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378872_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395579_at | - | - |
| Rat Genome 230 2.0 Array | 1378872_at | - | - |
| Affymetrix HT Rat Focus Array | 1395579_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395579_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100053727.rn.1 | TC0100006817.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100053728.rn.1 | TC0100006817.rn.1 | RTA TC |
| Rat Expression Set 230 | 1395579_PM_at | - | - |
| Rat Expression Set 230 | 1395579_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726358 | - | - |
| Rat Genome U34 Set | rc_AI176609_at | - | - |
| Rat Genome U34 Set | rc_AI179535_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21661661 | - | - |

Dhx32
Genomic Map