Assay Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004205.1 | NP_001004205.1 | 1 - 2 | 573 | 103 | |
| GenBank mRNA | BC079132.1 | - | 1 - 2 | 573 | 7127390 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Rilpl2
Genomic Map