Assay Details
| Gene Symbol: Adrbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012897.2 | NP_037029.1 | 15 - 16 | 1330 | 91 | |
| XM_008769317.1 | XP_008767539.1 | 14 - 15 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Adrbk2
Genomic Map