Assay Details
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012897.2 | NP_037029.1 | 3 - 4 | 258 | 78 | |
XM_008769317.1 | XP_008767539.1 | 2 - 3 | 149 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
