Assay Details
| Gene Symbol: Ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025665.1 | NP_001020836.1 | 8 - 9 | 1376 | 120 | |
| GenBank mRNA | BC097409.1 | - | 8 - 9 | 1376 | 7454160 | 120 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P302043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96447 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P302043 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P302043 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P302043 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P302043 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34284 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41825 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48334 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10758623 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758616 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758619 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758622 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758620 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758621 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10758617 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10758614 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200005852.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005840.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005843.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005844.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005845.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005853.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005867.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005861.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005841.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005848.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005846.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005858.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005860.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200005864.rn.1 | TC1200000657.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10758614 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20861406 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369786 | - | - |

Ccdc63
Genomic Map