Assay Details
| Gene Symbol: Dcun1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134798.1 | NP_001128270.1 | 3 - 4 | 399 | 78 | |
| XM_008771407.1 | XP_008769629.1 | 3 - 4 | 483 | 78 | ||
| XM_008771408.1 | XP_008769630.1 | 3 - 4 | 484 | 78 | ||
| GenBank mRNA | FQ224263.1 | - | 3 - 4 | 413 | 78 | |
| Gene Symbol: NEWGENE_1582994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771413.1 | XP_008769635.1 | 3 - 4 | 567 | 78 | |
| XM_008771414.1 | XP_008769636.1 | 3 - 4 | 567 | 78 | ||
| XM_008771415.1 | XP_008769637.1 | 3 - 4 | 567 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Dcun1d2
Genomic Map