Assay Details
| Gene Symbol: Adam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014772.1 | NP_001014772.1 | 21 - 22 | 2474 | 63 | |
| GenBank mRNA | AY953138.1 | - | 21 - 22 | 2474 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
membrane protein ectodomain proteolysis
cell adhesion
cell-matrix adhesion
transforming growth factor beta receptor signaling pathway
response to manganese ion
keratinocyte differentiation
cell adhesion mediated by integrin
positive regulation of cell adhesion mediated by integrin
cell-cell adhesion mediated by integrin
positive regulation of macrophage fusion
response to tumor necrosis factor
response to laminar fluid shear stress
monocyte activation
response to hydrogen peroxide
positive regulation of protein secretion
positive regulation of membrane protein ectodomain proteolysis
PMA-inducible membrane protein ectodomain proteolysis
response to glucocorticoid
positive regulation of keratinocyte migration
response to calcium ion
cellular response to lipopolysaccharide
molecular_function
metalloendopeptidase activity
protein kinase C binding
integrin binding
collagen binding
metallopeptidase activity
zinc ion binding
SH3 domain binding
laminin binding
activation of MAPKK activity
membrane protein ectodomain proteolysis
cell adhesion
cell-matrix adhesion
transforming growth factor beta receptor signaling pathway
response to manganese ion
keratinocyte differentiation
cell adhesion mediated by integrin
positive regulation of cell adhesion mediated by integrin
cell-cell adhesion mediated by integrin
positive regulation of macrophage fusion
response to tumor necrosis factor
response to laminar fluid shear stress
monocyte activation
response to hydrogen peroxide
positive regulation of protein secretion
positive regulation of membrane protein ectodomain proteolysis
PMA-inducible membrane protein ectodomain proteolysis
response to glucocorticoid
positive regulation of keratinocyte migration
response to calcium ion
cellular response to lipopolysaccharide
molecular_function
metalloendopeptidase activity
protein kinase C binding
integrin binding
collagen binding
metallopeptidase activity
zinc ion binding
SH3 domain binding
laminin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P927439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95959 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P927439 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P927439 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P927439 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P283807 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P927439 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8973 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38447 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43871 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91449 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22125 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41191 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10788916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788907 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788918 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788910 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788908 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788911 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788917 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788914 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788905 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788912 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788904 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788915 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788909 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788913 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10788899 | - | - |
| Rat Genome 230 2.0 Array | 1375337_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375337_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600006324.rn.1 | TC1600000833.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375337_at | - | - |
| Rat Expression Set 230 | 1375337_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788899 | - | - |
| Rat Genome U34 Set | AA851120_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20855186 | - | - |

Adam9
Genomic Map