Assay Details
| Gene Symbol: Shcbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105961.2 | NP_001099431.1 | 1 - 2 | 375 | 60 | |
| GenBank mRNA | BC160910.1 | - | 1 - 2 | 375 | 7456157 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Shcbp1l
Genomic Map