Assay Details
| Gene Symbol: Tmco5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234788.2 | XP_006234850.1 | 6 - 7 | 555 | 75 | |
| XM_006234789.2 | XP_006234851.1 | 6 - 7 | 535 | 75 | ||
| XM_006234790.2 | XP_006234852.1 | 6 - 7 | 580 | 75 | ||
| XM_006234791.2 | XP_006234853.1 | 5 - 6 | 437 | 75 | ||
| XM_008762112.1 | XP_008760334.1 | 6 - 7 | 612 | 75 | ||
More Information
Set Membership: |
|

Tmco5a
Genomic Map